Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022198.2 | NP_071534.2 | 9 - 10 | 1644 | 104 |
XM_006256828.2 | XP_006256890.1 | 9 - 10 | 1947 | 104 | |
GenBank mRNA | BC092209.1 | - | 9 - 10 | 1644 | 104 |
FQ210965.1 | - | 9 - 10 | 1683 | 104 | |
Z36944.1 | - | 7 - 8 | 1352 | 104 |