Assay Design
Gene Transcripts
- Gene Symbol
- Arf1
- Entrez Gene ID
-
64310
- Gene Name
- ADP-ribosylation factor 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.10: 45562700 - 45579192 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_022518.3 |
NP_071963.1 |
1 - 2 |
40 |
67 |
GenBank mRNA |
L12380.1 |
- |
1 - 2 |
40 |
67 |
View Full Product Details