Assay Design



Gene Transcripts

Gene Symbol
Chrna10
Entrez Gene ID
64574
Gene Name
cholinergic receptor, nicotinic, alpha 10 (neuronal)
Gene Aliases
-
Chromosome Location
Chr.1: 167206722 - 167210919 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022639.1 NP_072161.1 4 - 5 955 77
XM_006229891.2 XP_006229953.1 3 - 4 1046 77
GenBank mRNA AF196344.1 - 4 - 5 955 77

View Full Product Details