Assay Design



Gene Transcripts

Gene Symbol
Hes1
Entrez Gene ID
29577
Gene Name
hes family bHLH transcription factor 1
Gene Aliases
-
Chromosome Location
Chr.11: 74312837 - 74315249 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024360.3 NP_077336.3 1 - 2 313 63
GenBank mRNA BC061730.1 - 1 - 2 313 63
D13417.1 - 1 - 2 351 63
L04527.1 - - 875 63

View Full Product Details