Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031031.2 | NP_112293.1 | 6 - 7 | 1034 | 73 |
GenBank mRNA | AY625271.1 | - | 4 - 5 | 622 | 73 |
BC081785.1 | - | 6 - 7 | 1034 | 73 | |
FQ212973.1 | - | 2 - 3 | 205 | 73 | |
U07971.1 | - | 6 - 7 | 1026 | 73 |