Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031546.1 | NP_113734.1 | 3 - 4 | 444 | 72 |
GenBank mRNA | AB037934.1 | - | 3 - 4 | 444 | 72 |
BC078794.1 | - | 3 - 4 | 439 | 72 | |
D38467.1 | - | 3 - 4 | 438 | 72 | |
FQ219778.1 | - | 3 - 4 | 447 | 72 | |
X69021.1 | - | 3 - 4 | 433 | 72 |