Assay Design



Gene Transcripts

Gene Symbol
Cyp4a8
Entrez Gene ID
266674
Gene Name
cytochrome P450, family 4, subfamily a, polypeptide 8
Gene Aliases
Cyp4a12
Chromosome Location
Chr.5: 133978953 - 134008255 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031605.2 NP_113793.2 3 - 4 389 149
GenBank mRNA BC081771.1 - 3 - 4 389 149
M37828.1 - 3 - 4 379 149

View Full Product Details