Assay Design
Gene Transcripts
- Gene Symbol
- Ctla4
- Entrez Gene ID
-
63835
- Gene Name
- cytotoxic T-lymphocyte-associated protein 4
- Gene Aliases
-
CTLA-4, Cd152, sCTLA4
- Chromosome Location
-
Chr.9: 67699397 - 67706130 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_031674.1 |
NP_113862.1 |
3 - 4 |
570 |
62 |
GenBank mRNA |
U37121.1 |
- |
3 - 4 |
570 |
62 |
View Full Product Details