Assay Design



Gene Transcripts

Gene Symbol
Nsfl1c
Entrez Gene ID
83809
Gene Name
NSFL1 (p97) cofactor (p47)
Gene Aliases
XY40, p47
Chromosome Location
Chr.3: 146980923 - 147005475 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031981.2 NP_114187.1 5 - 6 640 61
XM_006235254.2 XP_006235316.1 6 - 7 613 61
GenBank mRNA AB002086.1 - 5 - 6 640 61
BC072464.1 - 5 - 6 537 61
FQ228953.1 - 6 - 7 578 61
Y10769.1 - 5 - 6 537 61

View Full Product Details