Assay Design



Gene Transcripts

Gene Symbol
Gde1
Entrez Gene ID
60418
Gene Name
glycerophosphodiester phosphodiesterase 1
Gene Aliases
Mir16
Chromosome Location
Chr.1: 188367550 - 188407125 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032615.2 NP_116004.2 - 945 71
GenBank mRNA AF212861.1 - - 890 71
BC070897.1 - - 945 71

View Full Product Details