Assay Design
Gene Transcripts
- Gene Symbol
- Il12a
- Entrez Gene ID
-
84405
- Gene Name
- interleukin 12A
- Gene Aliases
-
-
- Chromosome Location
-
Chr.2: 165076945 - 165083996 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_053390.1 |
NP_445842.1 |
2 - 3 |
181 |
67 |
GenBank mRNA |
AF177031.1 |
- |
2 - 3 |
181 |
67 |
View Full Product Details