Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053527.1 | NP_445979.1 | 13 - 14 | 2002 | 116 |
GenBank mRNA | AF000578.2 | - | 13 - 14 | 2002 | 116 |
BC087007.1 | - | 14 - 15 | 2049 | 116 | |
BC161839.1 | - | 13 - 14 | 1966 | 116 |