Assay Design



Gene Transcripts

Gene Symbol
Klf15
Entrez Gene ID
85497
Gene Name
Kruppel-like factor 15
Gene Aliases
-
Chromosome Location
Chr.4: 122365092 - 122377690 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053536.1 NP_445988.1 2 - 3 257 72
XM_008763135.1 XP_008761357.1 1 - 2 394 72
GenBank mRNA AB020759.1 - 2 - 3 257 72

View Full Product Details