Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053634.1 | NP_446086.1 | 8 - 9 | 742 | 105 |
GenBank mRNA | AB036792.1 | - | 8 - 9 | 742 | 105 |
BC088123.1 | - | 8 - 9 | 734 | 105 | |
FQ220557.1 | - | 8 - 9 | 745 | 105 | |
FQ233041.1 | - | 8 - 9 | 745 | 105 |