Assay Design



Gene Transcripts

Gene Symbol
Lim2
Entrez Gene ID
114903
Gene Name
lens intrinsic membrane protein 2
Gene Aliases
MP20, Mp19
Chromosome Location
Chr.1: 98495082 - 98501248 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053771.1 NP_446223.1 2 - 3 216 58
GenBank mRNA L04191.1 - 2 - 3 194 58
M87053.1 - 2 - 3 216 58
S55224.1 - 2 - 3 207 58

View Full Product Details