Assay Design



Gene Transcripts

Gene Symbol
Ifngr1
Entrez Gene ID
116465
Gene Name
interferon gamma receptor 1
Gene Aliases
Ifngr
Chromosome Location
Chr.1: 15062380 - 15080815 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053783.1 NP_446235.1 1 - 2 164 80
GenBank mRNA AF201901.1 - 1 - 2 164 80
BC062039.1 - 1 - 2 106 80

View Full Product Details