Assay Design



Gene Transcripts

Gene Symbol
Cplx2
Entrez Gene ID
116657
Gene Name
complexin 2
Gene Aliases
-
Chromosome Location
Chr.17: 10753862 - 10829263 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053878.1 NP_446330.1 1 - 2 208 96
XM_006253596.2 XP_006253658.1 1 - 2 244 96
GenBank mRNA D70816.1 - 1 - 2 208 96

View Full Product Details