Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053907.1 | NP_446359.1 | 5 - 6 | 762 | 105 |
GenBank mRNA | AF039852.1 | - | 5 - 6 | 712 | 105 |
BC088122.1 | - | 5 - 6 | 760 | 105 | |
FQ226037.1 | - | 5 - 6 | 589 | 105 | |
FQ230283.1 | - | 5 - 6 | 589 | 105 | |
FQ230990.1 | - | 5 - 6 | 589 | 105 | |
FQ231646.1 | - | 5 - 6 | 588 | 105 | |
FQ231744.1 | - | 5 - 6 | 588 | 105 | |
FQ231870.1 | - | 5 - 6 | 592 | 105 | |
FQ231976.1 | - | 5 - 6 | 645 | 105 | |
FQ233228.1 | - | 5 - 6 | 645 | 105 | |
FQ234709.1 | - | 5 - 6 | 645 | 105 | |
U75689.1 | - | 5 - 6 | 762 | 105 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_008770646.1 | XP_008768868.1 | 3 - 4 | 497 | 105 |