Assay Design



Gene Transcripts

Gene Symbol
Eif2b4
Entrez Gene ID
117019
Gene Name
eukaryotic translation initiation factor 2B, subunit 4 delta
Gene Aliases
Eif2b
Chromosome Location
Chr.6: 26546917 - 26552474 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053950.1 NP_446402.1 9 - 10 890 66
GenBank mRNA FQ233756.1 - 4 - 5 362 66
Z48225.1 - 9 - 10 890 66

View Full Product Details