Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053984.2 | NP_446436.1 | 1 - 1 | 1453 | 79 |
XM_008763985.1 | XP_008762207.1 | 2 - 2 | 79 | ||
XM_008763986.1 | XP_008762208.1 | 1 - 1 | 79 | ||
GenBank mRNA | X76168.1 | - | 1 - 1 | 1453 | 79 |