Assay Design
Gene Transcripts
- Gene Symbol
- Sv2b
- Entrez Gene ID
-
117556
- Gene Name
- synaptic vesicle glycoprotein 2b
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 136664201 - 136842679 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057207.3 |
NP_476555.1 |
1 - 2 |
59 |
60 |
GenBank mRNA |
L10362.1 |
- |
1 - 2 |
59 |
60 |
View Full Product Details