Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_131911.2 | NP_571986.1 | 2 - 3 | 379 | 68 |
XM_008763695.1 | XP_008761917.1 | 3 - 4 | 691 | 68 | |
GenBank mRNA | AB025581.1 | - | 2 - 3 | 216 | 68 |
BC086508.1 | - | 2 - 3 | 379 | 68 |