Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133311.1 | NP_579845.1 | 1 - 2 | 211 | 72 |
XM_008769421.1 | XP_008767643.1 | 1 - 2 | 363 | 72 | |
GenBank mRNA | AF004774.2 | - | 1 - 2 | 160 | 72 |
AF269251.1 | - | 1 - 2 | 211 | 72 |