Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_134401.2 | NP_599228.2 | 11 - 12 | 1575 | 70 |
XM_006231385.2 | XP_006231447.1 | 11 - 12 | 1637 | 70 | |
GenBank mRNA | BC089944.1 | - | 9 - 10 | 1106 | 70 |
U78304.1 | - | 2 - 3 | 216 | 70 |