Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138502.2 | NP_612511.1 | 2 - 3 | 245 | 78 |
XM_008763053.1 | XP_008761275.1 | 2 - 3 | 903 | 78 | |
GenBank mRNA | AY081195.1 | - | 1 - 2 | 128 | 78 |
BC107920.1 | - | 2 - 3 | 245 | 78 | |
FQ227013.1 | - | 2 - 3 | 871 | 78 |