Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138514.1 | NP_612523.1 | 4 - 5 | 653 | 85 |
GenBank mRNA | BC088105.1 | - | 4 - 5 | 675 | 85 |
BC098903.1 | - | 4 - 5 | 654 | 85 | |
J02861.1 | - | 4 - 5 | 655 | 85 | |
M32277.1 | - | 4 - 5 | 653 | 85 | |
M33994.1 | - | 4 - 5 | 642 | 85 |