Assay Design



Gene Transcripts

Gene Symbol
C1s
Entrez Gene ID
192262
Gene Name
complement component 1, s subcomponent
Gene Aliases
r-gsp
Chromosome Location
Chr.4: 157143592 - 157155592 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138900.1 NP_620255.1 4 - 5 656 69
GenBank mRNA BC062042.1 - 4 - 5 623 69
D88250.1 - 4 - 5 656 69

View Full Product Details