Assay Design



Gene Transcripts

Gene Symbol
Kcnc2
Entrez Gene ID
246153
Gene Name
potassium channel, voltage gated Shaw-related subfamily C, member 2
Gene Aliases
KShIIIA, Kv3.2
Chromosome Location
Chr.7: 54978786 - 55159362 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139217.1 NP_631963.1 3 - 4 1812 72
XM_006241327.2 XP_006241389.1 4 - 5 2064 72
GenBank mRNA M59211.1 - 3 - 4 1812 72
M84203.1 - 3 - 4 1855 72

View Full Product Details