Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144759.2 | NP_653360.2 | 5 - 6 | 578 | 82 |
XM_006248991.2 | XP_006249053.1 | 5 - 6 | 1364 | 82 | |
XM_006248992.1 | XP_006249054.1 | 4 - 5 | 409 | 82 | |
GenBank mRNA | EU741665.1 | - | 5 - 6 | 558 | 82 |