Assay Design
Gene Transcripts
- Gene Symbol
- Drgx
- Entrez Gene ID
-
252880
- Gene Name
- dorsal root ganglia homeobox
- Gene Aliases
-
Drg11, Prrxl1
- Chromosome Location
-
Chr.16: 8823872 - 8853430 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_145767.1 |
NP_665710.1 |
4 - 5 |
507 |
77 |
GenBank mRNA |
U29174.1 |
- |
4 - 5 |
507 |
77 |
View Full Product Details