Assay Design



Gene Transcripts

Gene Symbol
Rab38
Entrez Gene ID
252916
Gene Name
RAB38, member RAS oncogene family
Gene Aliases
R, Ruby
Chromosome Location
Chr.1: 152072716 - 152153449 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145774.2 NP_665717.1 2 - 3 528 72
GenBank mRNA BC070513.1 - 2 - 3 538 72
M94043.1 - 2 - 3 531 72

View Full Product Details