Assay Design
Gene Transcripts
- Gene Symbol
- Slc16a2
- Entrez Gene ID
-
259248
- Gene Name
- solute carrier family 16, member 2 (thyroid hormone transporter)
- Gene Aliases
-
-
- Chromosome Location
-
Chr.X: 74578600 - 74706068 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_147216.1 |
NP_671749.1 |
1 - 2 |
444 |
126 |
GenBank mRNA |
AJ496570.1 |
- |
1 - 2 |
444 |
126 |
View Full Product Details