Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172041.2 | NP_742038.2 | 5 - 6 | 876 | 75 |
NM_172042.1 | NP_742039.1 | 5 - 6 | 864 | 75 | |
XM_006250433.2 | XP_006250495.1 | 5 - 6 | 1087 | 75 | |
XM_006250434.2 | XP_006250496.1 | 5 - 6 | 1466 | 75 | |
GenBank mRNA | AF325671.1 | - | 5 - 6 | 864 | 75 |
AF385402.1 | - | 5 - 6 | 971 | 75 | |
AY555072.1 | - | 5 - 6 | 827 | 75 | |
AY695826.1 | - | 5 - 6 | 821 | 75 | |
AY727922.1 | - | 5 - 6 | 819 | 75 | |
DQ403851.1 | - | 5 - 6 | 876 | 75 |