Assay Design



Gene Transcripts

Gene Symbol
Prpmp5
Entrez Gene ID
257651
Gene Name
proline-rich protein MP5
Gene Aliases
Prb1, Prpg2
Chromosome Location
Chr.4: 166013226 - 166016335 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172065.2 NP_742062.1 4 - 5 933 84
GenBank mRNA L17318.1 - 4 - 5 934 84

View Full Product Details