Assay Design



Gene Transcripts

Gene Symbol
Igf2bp1
Entrez Gene ID
303477
Gene Name
insulin-like growth factor 2 mRNA binding protein 1
Gene Aliases
Imp1, ZBP1
Chromosome Location
Chr.10: 83774178 - 83823317 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175594.2 NP_783184.1 14 - 15 1867 87
XM_006247193.2 XP_006247255.1 14 - 15 2185 87
GenBank mRNA AF541940.1 - 14 - 15 1867 87
BC161831.1 - 14 - 15 1899 87

View Full Product Details