Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175598.2 | NP_783188.1 | 4 - 5 | 917 | 78 |
XM_006241180.2 | XP_006241242.1 | 4 - 5 | 1532 | 78 | |
XM_006241182.2 | XP_006241244.1 | 4 - 5 | 1532 | 78 | |
XM_008765234.1 | XP_008763456.1 | 4 - 5 | 1532 | 78 | |
XM_008765236.1 | XP_008763458.1 | 4 - 5 | 1532 | 78 | |
XM_008765237.1 | XP_008763459.1 | 4 - 5 | 1531 | 78 | |
GenBank mRNA | AY177755.1 | - | 4 - 5 | 917 | 78 |