Assay Design
Gene Transcripts
- Gene Symbol
- Gars
- Entrez Gene ID
-
297113
- Gene Name
- glycyl-tRNA synthetase
- Gene Aliases
-
GlyRS, RGD1559871
- Chromosome Location
-
Chr.4: 85235128 - 85276085 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC088347.1 |
- |
4 - 5 |
349 |
91 |
View Full Product Details