Assay Design



Gene Transcripts

Gene Symbol
Agtrap
Entrez Gene ID
298646
Gene Name
angiotensin II receptor-associated protein
Gene Aliases
-
Chromosome Location
Chr.5: 164888069 - 164898387 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007654.1 NP_001007655.1 1 - 2 64 62
GenBank mRNA BC082019.1 - 1 - 2 64 62

View Full Product Details