Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017198.1 | NP_058894.1 | 3 - 4 | 676 | 60 |
XM_006229752.1 | XP_006229814.1 | 3 - 4 | 527 | 60 | |
XM_006229753.1 | XP_006229815.1 | 3 - 4 | 524 | 60 | |
XM_006229754.2 | XP_006229816.1 | 3 - 4 | 516 | 60 | |
GenBank mRNA | U23443.1 | - | 3 - 4 | 482 | 60 |
U49953.1 | - | 3 - 4 | 676 | 60 |