Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021593.1 | NP_067604.1 | 7 - 8 | 660 | 92 |
XM_006250325.2 | XP_006250387.1 | 7 - 8 | 825 | 92 | |
XM_006250327.2 | XP_006250389.1 | 6 - 7 | 730 | 92 | |
XM_008769794.1 | XP_008768016.1 | 5 - 6 | 730 | 92 | |
XM_008769795.1 | XP_008768017.1 | 2 - 3 | 592 | 92 | |
GenBank mRNA | AF056031.1 | - | 7 - 8 | 660 | 92 |
BC088142.1 | - | 7 - 8 | 641 | 92 | |
FQ210049.1 | - | 7 - 8 | 658 | 92 |