Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031139.1 | NP_112401.1 | 8 - 9 | 1332 | 112 |
GenBank mRNA | AB035647.1 | - | 8 - 9 | 821 | 112 |
AB035648.1 | - | 8 - 9 | 740 | 112 | |
AB035649.1 | - | 7 - 8 | 620 | 112 | |
AB035650.1 | - | 7 - 8 | 774 | 112 | |
AB035651.1 | - | 5 - 6 | 454 | 112 | |
AB047556.1 | - | 8 - 9 | 1332 | 112 | |
BC128735.1 | - | 8 - 9 | 826 | 112 | |
X90823.1 | - | 6 - 7 | 656 | 112 |