Assay Design



Gene Transcripts

Gene Symbol
Cib1
Entrez Gene ID
81823
Gene Name
calcium and integrin binding 1 (calmyrin)
Gene Aliases
Cib, Sip2-28
Chromosome Location
Chr.1: 142014962 - 142020461 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031145.1 NP_112407.1 5 - 6 506 78
GenBank mRNA AF136585.1 - 5 - 6 506 78
BC091143.1 - 5 - 6 492 78

View Full Product Details