Assay Design



Gene Transcripts

Gene Symbol
Pafah1b1
Entrez Gene ID
83572
Gene Name
platelet-activating factor acetylhydrolase 1b, regulatory subunit 1
Gene Aliases
LIS1
Chromosome Location
Chr.10: 61457216 - 61498601 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031763.3 NP_113951.1 - 520 99
GenBank mRNA AF016049.1 - - 335 99
BC072510.1 - - 520 99
FQ224997.1 - - 244 99

View Full Product Details