Assay Design



Gene Transcripts

Gene Symbol
Spa17
Entrez Gene ID
85244
Gene Name
sperm autoantigenic protein 17
Gene Aliases
Sp17
Chromosome Location
Chr.8: 40066954 - 40078157 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053482.1 NP_445934.1 2 - 3 226 86
XM_006242812.2 XP_006242874.1 3 - 4 402 86
GenBank mRNA AJ131888.1 - 2 - 3 226 86

View Full Product Details