Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053573.1 | NP_446025.1 | 3 - 4 | 753 | 78 |
XM_006233875.2 | XP_006233937.1 | 3 - 4 | 713 | 78 | |
GenBank mRNA | U03416.1 | - | 3 - 4 | 615 | 78 |
U03417.1 | - | 3 - 4 | 753 | 78 |