Assay Design
Gene Transcripts
- Gene Symbol
- Cds2
- Entrez Gene ID
-
114101
- Gene Name
- CDP-diacylglycerol synthase (phosphatidate cytidylyltransferase) 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.3: 124896657 - 124935221 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_053643.1 |
NP_446095.1 |
6 - 7 |
663 |
93 |
GenBank mRNA |
AB052898.1 |
- |
6 - 7 |
663 |
93 |
View Full Product Details