Assay Design



Gene Transcripts

Gene Symbol
Kcnj9
Entrez Gene ID
116560
Gene Name
potassium channel, inwardly rectifying subfamily J, member 9
Gene Aliases
Girk3, Kir3.3
Chromosome Location
Chr.13: 90701432 - 90710148 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053834.1 NP_446286.1 3 - 4 1096 68
XM_008769730.1 XP_008767952.1 1 - 2 2058 68
GenBank mRNA L77929.1 - 3 - 4 1096 68

View Full Product Details