Assay Design



Gene Transcripts

Gene Symbol
Aldh9a1
Entrez Gene ID
64040
Gene Name
aldehyde dehydrogenase 9 family, member A1
Gene Aliases
Tmaba-dh
Chromosome Location
Chr.13: 85580828 - 85597497 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022273.2 NP_071609.2 4 - 5 591 61
GenBank mRNA AF170918.1 - 4 - 5 578 61
BC074019.1 - 4 - 5 591 61

View Full Product Details