Assay Design



Gene Transcripts

Gene Symbol
Adcy5
Entrez Gene ID
64532
Gene Name
adenylate cyclase 5
Gene Aliases
-
Chromosome Location
Chr.11: 68695839 - 68842493 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022600.1 NP_072122.1 10 - 11 2693 60
XM_008768744.1 XP_008766966.1 10 - 11 2866 60
GenBank mRNA M96159.1 - 10 - 11 2693 60

View Full Product Details