Assay Design



Gene Transcripts

Gene Symbol
Il12b
Entrez Gene ID
64546
Gene Name
interleukin 12B
Gene Aliases
Il12
Chromosome Location
Chr.10: 30034453 - 30048120 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022611.1 NP_072133.1 2 - 3 347 87
XM_006246147.2 XP_006246209.1 3 - 4 556 87
GenBank mRNA AF133197.1 - 2 - 3 347 87

View Full Product Details